siRNA Details
Gene Symbol: PHC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004426.2 | NP_004417.2 | 1 | 249 | ||
XM_005253334.2 | XP_005253391.1 | 2 | 273 | |||
XM_011520599.2 | XP_011518901.1 | 2 | 225 | |||
XM_011520600.2 | XP_011518902.1 | 2 | 225 | |||
XM_011520603.2 | XP_011518905.1 | 2 | 305 | |||
XM_017018955.1 | XP_016874444.1 | 2 | 593 | |||
XM_017018956.1 | XP_016874445.1 | 2 | 225 | |||
XM_017018957.1 | XP_016874446.1 | 2 | 225 | |||
XM_017018958.1 | XP_016874447.1 | 2 | 225 | |||
GenBank mRNA | BC157850.1 | - | 1 | 155 | 9057069 | |
CN388283.1 | - | 1 | 147 | |||
U89277.1 | - | 2 | 228 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
