siRNA Details
Gene Symbol: RNF144B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182757.3 | NP_877434.2 | 1 | 141 | ||
XM_005248986.3 | XP_005249043.1 | 1 | 162 | |||
GenBank mRNA | AB076367.1 | - | 1 | 128 | ||
AK096832.1 | - | 1 | 141 | |||
BC063311.1 | - | 1 | 15 | 30331343 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
