siRNA Details
Gene Symbol: TSPAN11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080509.2 | NP_001073978.1 | 6 | 616 | ||
XM_011520679.2 | XP_011518981.1 | 6 | 878 | |||
XM_011520680.2 | XP_011518982.1 | 6 | 856 | |||
GenBank mRNA | AK293947.1 | - | 6 | 563 | ||
AK300920.1 | - | 5 | 521 | |||
BC127660.1 | - | 4 | 382 | |||
BC127661.1 | - | 4 | 382 | |||
BC146917.1 | - | 6 | 562 | 9056845 | ||
BC146922.1 | - | 6 | 562 | 9056850 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
