siRNA Details
Gene Symbol: CD33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001082618.1 | NP_001076087.1 | 6 | 698 | ||
NM_001772.3 | NP_001763.3 | 7 | 1079 | |||
XM_011527531.2 | XP_011525833.1 | 13 | 2153 | |||
XM_011527532.2 | XP_011525834.1 | 12 | 2116 | |||
XM_017027508.1 | XP_016882997.1 | 13 | 2289 | |||
XM_017027509.1 | XP_016882998.1 | 11 | 2193 | |||
XM_017027510.1 | XP_016882999.1 | 11 | 1860 | |||
GenBank mRNA | AK090470.1 | - | 4 | 4105 | ||
AY162464.1 | - | 6 | 666 | |||
BC028152.1 | - | 7 | 1044 | 5217182 | ||
M23197.1 | - | 7 | 1051 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
