siRNA Details
Gene Symbol: ZNF614 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025040.3 | NP_079316.2 | 4 | 609 | ||
GenBank mRNA | AK097156.1 | - | 4 | 593 | ||
BC004930.1 | - | 4 | 531 | |||
BC101392.1 | - | 4 | 447 | 40026900 | ||
BC101393.2 | - | 4 | 447 | 40026901 | ||
BC101394.1 | - | 4 | 447 | 40026907 | ||
BC101395.1 | - | 4 | 447 | 40026909 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
