siRNA Details
Gene Symbol: VPS8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009921.2 | NP_001009921.1 | 23 | 2030 | ||
NM_015303.3 | NP_056118.2 | 22 | 2024 | |||
XM_005247251.3 | XP_005247308.1 | 23 | 2298 | |||
XM_005247253.4 | XP_005247310.1 | 23 | 2036 | |||
XM_006713556.3 | XP_006713619.1 | 15 | 1290 | |||
XM_011512599.2 | XP_011510901.1 | 10 | 796 | |||
XM_011512600.2 | XP_011510902.1 | 16 | 1317 | |||
XM_011512601.2 | XP_011510903.1 | 16 | 1497 | |||
XM_017006040.1 | XP_016861529.1 | 21 | 1871 | |||
XM_017006041.1 | XP_016861530.1 | 23 | 2036 | |||
GenBank mRNA | AB018347.1 | - | 15 | 1209 | ||
AK022945.1 | - | 1 | 39 | |||
AK056661.1 | - | 23 | 2002 | |||
AK292423.1 | - | 22 | 1996 | |||
AK309244.1 | - | 2 | 266 | |||
AL833838.1 | - | 20 | 1563 | |||
BC140768.1 | - | 23 | 1966 | 9021636 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
