siRNA Details
Gene Symbol: FLYWCH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142499.1 | NP_001135971.1 | 2 | 121 | ||
NM_001142500.1 | NP_001135972.1 | 2 | 211 | |||
NM_138439.2 | NP_612448.1 | 2 | 215 | |||
GenBank mRNA | AL542248.3 | - | 2 | 103 | ||
BC014089.2 | - | 2 | 93 | 4330613 | ||
BG120460.1 | - | 2 | 213 | |||
BI825861.1 | - | 2 | 176 | |||
DB455461.1 | - | 2 | 195 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
