siRNA Details
Gene Symbol: CR1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175710.1 | NP_783641.1 | 5 | 803 | ||
GenBank mRNA | AY114160.1 | - | 4 | 514 | ||
BC109190.1 | - | 6 | 892 | |||
BC109191.1 | - | 5 | 742 |
Gene Symbol: CR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000573.3 | NP_000564.2 | 5 | 846 | ||
NM_000651.4 | NP_000642.3 | 5 | 846 | |||
XM_006711166.2 | XP_006711229.1 | 5 | 822 | |||
XM_011509205.1 | XP_011507507.1 | 5 | 822 | |||
GenBank mRNA | AK309342.1 | - | 5 | 827 | ||
BC032550.1 | - | 5 | 735 | |||
X05309.1 | - | 4 | 552 | |||
X14362.1 | - | 5 | 633 | |||
Y00816.1 | - | 733 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
