siRNA Details
Gene Symbol: ANKRD30BL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027019.1 | 1 | 91 | |||
GenBank mRNA | BC057764.1 | - | 1 | 94 | ||
BG719107.1 | - | 1 | 99 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
