siRNA Details
Gene Symbol: GPX8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008397.3 | NP_001008398.2 | 3 | 2035 | ||
NM_001306197.1 | NP_001293126.1 | 3 | 1882 | |||
NM_001306198.1 | NP_001293127.1 | 2 | 1773 | |||
NM_001306201.1 | NP_001293130.1 | 3 | 1756 | |||
NR_131336.1 | 4 | 1949 | ||||
NR_131337.1 | 4 | 1891 | ||||
NR_131338.1 | 3 | 1687 | ||||
NR_131340.1 | 3 | 2025 | ||||
XM_006714631.2 | XP_006714694.1 | 3 | 2001 | |||
GenBank mRNA | AK022110.1 | - | 1 | 50 | ||
BX537894.1 | - | 4 | 1899 | |||
BX647770.1 | - | 2 | 1723 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
