siRNA Details
Gene Symbol: CNTNAP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033655.3 | NP_387504.2 | 15 | 2561 | ||
GenBank mRNA | AB051501.2 | - | 15 | 2468 | ||
BC132737.1 | - | 15 | 2463 | 40146759 | ||
BC144130.1 | - | 13 | 2184 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
