siRNA Details
Gene Symbol: C6orf25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025260.3 | NP_079536.2 | 2 | 282 | ||
NM_138272.2 | NP_612116.1 | 2 | 282 | |||
NM_138273.2 | NP_612117.1 | 2 | 282 | |||
NM_138274.2 | NP_612118.1 | 2 | 282 | |||
NM_138275.2 | NP_612119.1 | 2 | 282 | |||
NM_138277.2 | NP_612121.1 | 2 | 282 | |||
XM_011514920.2 | XP_011513222.1 | 2 | 316 | |||
XM_011514921.2 | XP_011513223.1 | 2 | 316 | |||
XM_017011330.1 | XP_016866819.1 | 2 | 316 | |||
XM_017011331.1 | XP_016866820.1 | 2 | 554 | |||
XM_017011332.1 | XP_016866821.1 | 2 | 316 | |||
XM_017011333.1 | XP_016866822.1 | 2 | 567 | |||
GenBank mRNA | AJ292259.1 | - | 2 | 242 | ||
AJ292260.1 | - | 2 | 242 | |||
AJ292261.1 | - | 2 | 242 | |||
AJ292262.1 | - | 2 | 242 | |||
AJ292263.1 | - | 2 | 242 | |||
AJ292264.1 | - | 2 | 242 | |||
AJ292265.1 | - | 2 | 242 | |||
AK092392.1 | - | 2 | 253 | |||
BC113719.1 | - | 2 | 282 | 8322771 | ||
BC113721.1 | - | 2 | 282 | 8322773 | ||
BC143850.1 | - | 2 | 282 | 9052366 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
