siRNA Details
Gene Symbol: ATP23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320408.1 | NP_001307337.1 | 4 | 657 | ||
NM_001320409.1 | NP_001307338.1 | 3 | 571 | |||
NM_001320410.1 | NP_001307339.1 | 5 | 788 | |||
NM_033276.3 | NP_150592.1 | 4 | 617 | |||
XM_011538976.2 | XP_011537278.1 | 3 | 292 | |||
XM_017020206.1 | XP_016875695.1 | 4 | 407 | |||
GenBank mRNA | AF078164.2 | - | 4 | 547 | ||
BC012776.2 | - | 4 | 459 | 3936722 | ||
BC115381.1 | - | 4 | 458 | 40036852 | ||
BC115382.1 | - | 4 | 457 | 40036859 | ||
BM544500.1 | - | 4 | 616 | |||
BM557436.1 | - | 5 | 622 | |||
BU601924.1 | - | 4 | 620 | |||
DA711361.1 | - | 3 | 534 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
