siRNA Details
Gene Symbol: HINT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005340.6 | NP_005331.1 | 3 | 564 | ||
NR_024610.2 | 4 | 753 | ||||
NR_024611.2 | 3 | 599 | ||||
NR_073488.1 | 5 | 944 | ||||
NR_134494.1 | 5 | 909 | ||||
NR_134495.1 | 4 | 1013 | ||||
GenBank mRNA | AA076103.1 | - | 1 | 95 | ||
AK026557.1 | - | 3 | 473 | |||
AK054976.1 | - | 1 | 2556 | |||
AK056902.1 | - | 1 | 1895 | |||
BC001287.1 | - | 3 | 443 | 3458182 | ||
BC007090.1 | - | 3 | 516 | 4250172 | ||
BG527380.1 | - | 4 | 658 | |||
BG708765.1 | - | 3 | 569 | |||
CN290233.1 | - | 3 | 416 | |||
CN290239.1 | - | 3 | 541 | |||
DB125655.1 | - | 3 | 551 | |||
U27143.1 | - | 3 | 464 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
