siRNA Details
Gene Symbol: GTPBP8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014170.2 | NP_054889.2 | 1 | 288 | ||
NM_138485.1 | NP_612494.1 | 1 | 288 | |||
GenBank mRNA | AF161484.1 | - | 1 | 258 | ||
AK289611.1 | - | 1 | 250 | |||
AK292651.1 | - | 1 | 307 | |||
BC010137.2 | - | 1 | 254 | |||
BC037163.1 | - | 1 | 291 | 4620206 | ||
BI830308.1 | - | 1 | 288 | |||
BX354218.2 | - | 1 | 256 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
