siRNA Details
Gene Symbol: SLC9C1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320531.1 | NP_001307460.1 | 23, 24 | 3165 | ||
NM_183061.2 | NP_898884.1 | 24, 25 | 3309 | |||
NR_135297.1 | 18, 19 | 2368 | ||||
XM_006713603.2 | XP_006713666.1 | 24, 25 | 3344 | |||
XM_011512718.1 | XP_011511020.1 | 23, 24 | 3147 | |||
XM_011512719.1 | XP_011511021.1 | 23, 24 | 3138 | |||
XM_011512721.1 | XP_011511023.1 | 25, 26 | 3487 | |||
XM_011512725.2 | XP_011511027.1 | 14, 15 | 1858 | |||
XM_017006246.1 | XP_016861735.1 | 22, 23 | 3012 | |||
XM_017006247.1 | XP_016861736.1 | 23, 24 | 3123 | |||
XM_017006248.1 | XP_016861737.1 | 21, 22 | 2913 | |||
GenBank mRNA | AK128084.1 | - | 24, 25 | 3309 | ||
BC044801.1 | - | 23, 24 | 3134 | 4827529 | ||
BC143774.1 | - | 17, 18 | 2213 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
