siRNA Details
Gene Symbol: SLC22A25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199352.3 | NP_955384.3 | 1 | 238 | ||
XM_017017684.1 | XP_016873173.1 | 4 | 1317 | |||
XM_017017685.1 | XP_016873174.1 | 5 | 1445 | |||
XM_017017686.1 | XP_016873175.1 | 4 | 1753 | |||
XM_017017687.1 | XP_016873176.1 | 4 | 1858 | |||
XM_017017695.1 | XP_016873184.1 | 4 | 1858 | |||
GenBank mRNA | AK127610.1 | - | 1 | 113 | ||
AY437532.1 | - | 1 | 238 | |||
BC101315.1 | - | 1 | 233 | |||
BC101316.1 | - | 1 | 238 | |||
BC101317.1 | - | 1 | 238 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
