siRNA Details
Gene Symbol: FAM53A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013622.3 | NP_001013644.1 | 4 | 446 | ||
NM_001174070.2 | NP_001167541.1 | 4 | 493 | |||
NM_001297435.1 | NP_001284364.1 | 4 | 846 | |||
XM_005247948.3 | XP_005248005.1 | 4 | 1143 | |||
XM_011513402.2 | XP_011511704.1 | 4 | 1149 | |||
XM_011513403.1 | XP_011511705.1 | 4 | 1143 | |||
XM_011513404.2 | XP_011511706.1 | 4 | 1143 | |||
XM_011513405.1 | XP_011511707.1 | 4 | 1143 | |||
XM_017007802.1 | XP_016863291.1 | 4 | 1143 | |||
GenBank mRNA | AK125566.1 | - | 4 | 441 | ||
BC070112.1 | - | 4 | 446 | 5268061 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
