siRNA Details
Gene Symbol: CNOT6L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286790.1 | NP_001273719.1 | 11 | 1424 | ||
NM_144571.2 | NP_653172.2 | 11 | 1353 | |||
XM_011531806.2 | XP_011530108.1 | 12 | 1488 | |||
XM_011531807.2 | XP_011530109.1 | 12 | 1462 | |||
XM_011531808.2 | XP_011530110.1 | 11 | 1434 | |||
XM_011531809.1 | XP_011530111.1 | 11 | 1379 | |||
XM_011531810.1 | XP_011530112.1 | 11 | 1939 | |||
XM_017007956.1 | XP_016863445.1 | 11 | 1376 | |||
XM_017007957.1 | XP_016863446.1 | 11 | 1702 | |||
GenBank mRNA | AK027214.1 | - | 2 | 42 | ||
AK058188.1 | - | 11 | 1250 | |||
AK303417.1 | - | 11 | 1402 | |||
AL133112.1 | - | 5 | 651 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
