siRNA Details
Gene Symbol: LGALS9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002308.3 | NP_002299.2 | 10 | 1323 | ||
NM_009587.2 | NP_033665.1 | 11 | 1419 | |||
NR_024043.1 | 10 | 1252 | ||||
XM_006721892.2 | XP_006721955.1 | 9 | 1250 | |||
XM_006721893.2 | XP_006721956.1 | 10 | 1248 | |||
XM_006721894.3 | XP_006721957.1 | 9 | 1123 | |||
XM_006721895.3 | XP_006721958.1 | 8 | 1087 | |||
XM_017024623.1 | XP_016880112.1 | 10 | 1375 | |||
GenBank mRNA | AB005894.1 | - | 10 | 1264 | ||
AB006782.1 | - | 11 | 1382 | |||
AK126017.1 | - | 10 | 2673 | |||
AK223232.1 | - | 10 | 1287 | |||
AK301624.1 | - | 10 | 1126 | |||
BC110340.1 | - | 10 | 1264 | 5760999 | ||
Z49107.1 | - | 10 | 1292 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
