siRNA Details
Gene Symbol: PRR13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005354.2 | NP_001005354.1 | 3 | 238 | ||
NM_018457.3 | NP_060927.1 | 3 | 388 | |||
GenBank mRNA | AF217517.1 | - | 1 | 301 | ||
AK074398.1 | - | 1 | 284 | |||
BC001464.2 | - | 3 | 374 | |||
BC014257.2 | - | 1 | 288 | 4765821 | ||
BC016064.1 | - | 1 | 285 | 4852862 | ||
BC021019.1 | - | 1 | 256 | |||
BC071979.1 | - | 1 | 231 | |||
BX640962.1 | - | 2 | 126 | |||
CN392914.1 | - | 3 | 402 | |||
CR533459.1 | - | 2 | 224 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
