siRNA Details
Gene Symbol: FAM133B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040057.2 | NP_001035146.1 | 12 | 1195 | ||
NM_001288584.1 | NP_001275513.1 | 11 | 1180 | |||
NM_152789.3 | NP_690002.2 | 11 | 1096 | |||
NR_109929.1 | 11 | 1055 | ||||
XM_011516004.2 | XP_011514306.1 | 11 | 1359 | |||
XM_011516005.2 | XP_011514307.1 | 11 | 2680 | |||
GenBank mRNA | AK125498.1 | - | 9 | 1476 | ||
AL833918.2 | - | 12 | 1195 | |||
BC032461.1 | - | 11 | 1041 | 5197918 | ||
BC057771.1 | - | 12 | 1186 | 5301001 | ||
BC090868.1 | - | 11 | 1080 | 6190536 |
Gene Symbol: FAM133DP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_034169.1 | 1 | 1040 | |||
GenBank mRNA | T77355.1 | - | 1 | 154 |
Gene Symbol: LOC100421561 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC157872.1 | - | 1 | 1053 | 9057091 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
