siRNA Details
Gene Symbol: GIPC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133261.2 | NP_573568.1 | 4 | 658 | ||
XM_005259492.3 | XP_005259549.1 | 4 | 698 | |||
GenBank mRNA | AB073738.1 | - | 4 | 658 | ||
BC117312.1 | - | 4 | 656 | 40125863 | ||
BC117314.1 | - | 4 | 656 | 40125865 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
