siRNA Details
Gene Symbol: ANKS6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173551.4 | NP_775822.3 | 2 | 871 | ||
XM_005251793.4 | XP_005251850.1 | 2 | 858 | |||
XM_005251794.4 | XP_005251851.1 | 2 | 858 | |||
XM_006716998.3 | XP_006717061.1 | 2 | 858 | |||
XM_006716999.2 | XP_006717062.1 | 2 | 1943 | |||
XM_011518357.2 | XP_011516659.1 | 1 | 2819 | |||
XM_017014445.1 | XP_016869934.1 | 2 | 858 | |||
XM_017014446.1 | XP_016869935.1 | 1 | 2819 | |||
GenBank mRNA | AK127702.1 | - | 1 | 458 | ||
BC012981.2 | - | 1 | 1834 | |||
BX462630.2 | - | 2 | 867 | |||
BY994327.1 | - | 2 | 713 | |||
CR749467.1 | - | 2 | 542 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
