siRNA Details
Gene Symbol: USP45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080481.1 | NP_001073950.1 | 3 | 675 | ||
XM_005267169.3 | XP_005267226.1 | 3 | 278 | |||
XM_005267170.4 | XP_005267227.1 | 3 | 247 | |||
XM_005267172.3 | XP_005267229.1 | 3 | 247 | |||
XM_005267175.3 | XP_005267232.1 | 3 | 246 | |||
XM_011536200.2 | XP_011534502.1 | 3 | 1505 | |||
XM_017011382.1 | XP_016866871.1 | 3 | 672 | |||
XM_017011383.1 | XP_016866872.1 | 3 | 198 | |||
XM_017011384.1 | XP_016866873.1 | 3 | 175 | |||
XM_017011385.1 | XP_016866874.1 | 3 | 246 | |||
XM_017011386.1 | XP_016866875.1 | 3 | 246 | |||
GenBank mRNA | AJ583819.2 | - | 2 | 142 | ||
AK310643.1 | - | 3 | 179 | |||
AL713747.2 | - | 1 | 41 | |||
BC005991.1 | - | 3 | 193 | |||
BC150648.2 | - | 3 | 242 | 9057019 | ||
BC157838.1 | - | 3 | 242 | 9057057 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
