siRNA Details
Gene Symbol: TRIM49 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020358.2 | NP_065091.1 | 8 | 2067 | ||
GenBank mRNA | AB037682.1 | - | 8 | 2067 |
Gene Symbol: TRIM49C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195234.1 | NP_001182163.1 | 8 | 2067 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
