siRNA Details
Gene Symbol: TRIM49 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020358.2 | NP_065091.1 | 3 | 499 | ||
XM_017018027.1 | XP_016873516.1 | 1 | 174 | |||
GenBank mRNA | AB037682.1 | - | 3 | 499 | ||
BC126472.1 | - | 2 | 293 | 8992188 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
