siRNA Details
Gene Symbol: FAM132A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014980.2 | NP_001014980.1 | 1 | 124 | ||
XM_011541435.2 | XP_011539737.1 | 1 | 222 | |||
XM_011541436.2 | XP_011539738.1 | 1 | 222 | |||
GenBank mRNA | BC089443.1 | - | 1 | 124 | 30554756 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
