siRNA Details
Gene Symbol: RPS27L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015920.3 | NP_057004.1 | 2, 3 | 253 | ||
GenBank mRNA | AF070668.1 | - | 2, 3 | 175 | ||
AK024591.1 | - | 2, 3 | 188 | |||
BC003667.1 | - | 2, 3 | 185 | 3827362 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
