siRNA Details
Gene Symbol: LINC01547 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027128.1 | 3 | 711 | |||
NR_027129.1 | 4 | 855 | ||||
GenBank mRNA | AF380178.1 | - | 4 | 787 | ||
AY040088.1 | - | 5 | 737 | |||
BC128244.1 | - | 3 | 569 | |||
BC128245.1 | - | 3 | 569 | |||
BI829356.1 | - | 3 | 675 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
