siRNA Details
Gene Symbol: RESP18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007089.3 | NP_001007090.3 | 2, 3 | 227 | ||
GenBank mRNA | AF437883.3 | - | 2, 3 | 227 | ||
AF439399.1 | - | 1, 2 | 101 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
