siRNA Details
Gene Symbol: MYCL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033081.2 | NP_001028253.1 | 2 | 3556 | ||
NM_001033082.2 | NP_001028254.2 | 3 | 3192 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
