siRNA Details
Gene Symbol: AKR1C8P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027916.2 | 2 | 218 | |||
GenBank mRNA | AK131335.1 | - | 2 | 220 | ||
BC101204.1 | - | 2 | 212 | 40021857 | ||
BC101205.1 | - | 2 | 212 | 40021860 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
