siRNA Details
Gene Symbol: FSD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007122.3 | NP_001007123.1 | 13 | 5918 | ||
NM_001281805.1 | NP_001268734.1 | 13 | 5900 | |||
NM_001281806.1 | NP_001268735.1 | 12 | 5783 | |||
GenBank mRNA | AL833295.1 | - | 13 | 5917 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
