siRNA Details
Gene Symbol: SLC10A7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029998.5 | NP_001025169.1 | 3 | 548 | ||
NM_001300842.2 | NP_001287771.1 | 3 | 548 | |||
NM_001317816.1 | NP_001304745.1 | 3 | 548 | |||
NM_032128.4 | NP_115504.1 | 3 | 548 | |||
NR_133924.1 | 3 | 548 | ||||
XM_011532311.2 | XP_011530613.1 | 3 | 548 | |||
XM_017008689.1 | XP_016864178.1 | 3 | 548 | |||
XM_017008690.1 | XP_016864179.1 | 3 | 548 | |||
XM_017008691.1 | XP_016864180.1 | 3 | 548 | |||
GenBank mRNA | AK075364.1 | - | 1 | 60 | ||
AK291046.1 | - | 3 | 416 | |||
AL136728.1 | - | 3 | 476 | |||
AY346324.1 | - | 3 | 511 | |||
BC063471.1 | - | 3 | 461 | |||
BC120970.1 | - | 3 | 407 | |||
BC127626.1 | - | 3 | 407 | |||
BC127627.1 | - | 3 | 407 | |||
BC148252.1 | - | 3 | 407 | |||
BC150308.1 | - | 3 | 517 | 8860437 | ||
CR933647.1 | - | 3 | 542 | |||
CR936751.1 | - | 3 | 405 | |||
DQ122860.1 | - | 3 | 294 | |||
DQ122861.1 | - | 3 | 294 | |||
DQ871036.1 | - | 3 | 294 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
