siRNA Details
Gene Symbol: LRRC29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004055.1 | NP_001004055.1 | 1, 2 | 32 | ||
XM_017023126.1 | XP_016878615.1 | 1, 2 | 96 | |||
XM_017023130.1 | XP_016878619.1 | 1, 2 | 94 | |||
XM_017023131.1 | XP_016878620.1 | 1, 2 | 44 | |||
GenBank mRNA | AF176701.1 | - | 1, 2 | 41 | ||
AK316602.1 | - | 1, 2 | 45 | |||
BI085522.1 | - | 1, 2 | 32 |
Gene Symbol: LOC105369155 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK298088.1 | - | 1, 2 | 61 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
