siRNA Details
Gene Symbol: FAM90A10P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046351.1 | 3 | 531 | |||
GenBank mRNA | DC395395.1 | - | 3 | 532 |
Gene Symbol: FAM90A7P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046343.1 | 3 | 531 | |||
GenBank mRNA | AK301810.1 | - | 3 | 532 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
