siRNA Details
Gene Symbol: KIAA1324L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142749.2 | NP_001136221.1 | 4 | 745 | ||
NM_001291990.1 | NP_001278919.1 | 4 | 503 | |||
NM_001291991.1 | NP_001278920.1 | 2 | 186 | |||
NM_152748.3 | NP_689961.3 | 3 | 342 | |||
XM_006715894.3 | XP_006715957.1 | 4 | 445 | |||
XM_011515918.2 | XP_011514220.1 | 4 | 596 | |||
XM_011515921.1 | XP_011514223.1 | 3 | 618 | |||
XM_017011843.1 | XP_016867332.1 | 4 | 741 | |||
XM_017011844.1 | XP_016867333.1 | 3 | 335 | |||
GenBank mRNA | AK055902.1 | - | 2 | 167 | ||
AK296250.1 | - | 3 | 259 | |||
AK303619.1 | - | 4 | 505 | |||
BX346336.2 | - | 4 | 749 | |||
DA240191.1 | - | 4 | 533 | |||
DY654933.1 | - | 4 | 412 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
