siRNA Details
Gene Symbol: MED22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133640.4 | NP_598395.1 | 4 | 642 | ||
NM_181491.2 | NP_852468.1 | 4 | 642 | |||
GenBank mRNA | AJ224358.1 | - | 4 | 587 | ||
AJ224359.1 | - | 4 | 587 | |||
AK124518.1 | - | 4 | 954 | |||
AK126069.1 | - | 4 | 623 | |||
BC024225.2 | - | 4 | 585 | 3009749 | ||
BC040111.1 | - | 4 | 592 | 6056006 | ||
BI756582.1 | - | 4 | 603 | |||
X85178.1 | - | 4 | 593 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
