siRNA Details
Gene Symbol: ABHD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007011.7 | NP_008942.3 | 15 | 4130 | ||
NM_152924.4 | NP_690888.1 | 11 | 3732 | |||
XM_005254832.1 | XP_005254889.1 | 14 | 3768 | |||
XM_005254833.1 | XP_005254890.1 | 11 | 3376 | |||
XM_011521165.1 | XP_011519467.1 | 12 | 3581 | |||
GenBank mRNA | AK127559.1 | - | 1 | 124 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
