siRNA Details
Gene Symbol: CCDC73 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008391.3 | NP_001008392.2 | 13 | 1089 | ||
XM_011520139.2 | XP_011518441.1 | 12 | 950 | |||
GenBank mRNA | AK128159.1 | - | 4 | 308 | ||
AK131470.1 | - | 13 | 1072 | |||
BC062749.1 | - | 13 | 1059 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
