siRNA Details
Gene Symbol: TBL1Y | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033284.1 | NP_150600.1 | 1 | 104 | ||
NM_134258.1 | NP_599020.1 | 1 | 104 | |||
NM_134259.1 | NP_599021.1 | 1 | 104 | |||
XM_017030086.1 | XP_016885575.1 | 1 | 93 | |||
XM_017030087.1 | XP_016885576.1 | 1 | 93 | |||
GenBank mRNA | AF332220.1 | - | 1 | 104 | ||
AF332221.1 | - | 1 | 104 | |||
AF332222.1 | - | 1 | 104 | |||
AK056903.1 | - | 1 | 207 | |||
AK298210.1 | - | 1 | 93 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
