siRNA Details
Gene Symbol: FUNDC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023934.3 | NP_076423.2 | 5 | 906 | ||
GenBank mRNA | AF267862.1 | - | 1 | 936 | ||
AF320778.1 | - | 5 | 869 | |||
AJ272054.1 | - | 5 | 863 | |||
AK091092.1 | - | 2 | 2972 | |||
AK129832.1 | - | 1025 | ||||
BC000255.1 | - | 5 | 853 | 3355861 | ||
BC072685.1 | - | 5 | 811 | 6502640 | ||
BC108657.1 | - | 5 | 762 | 5752287 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
