siRNA Details
Gene Symbol: MAN2A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320977.1 | NP_001307906.1 | 25 | 6262 | ||
NM_006122.3 | NP_006113.2 | 23 | 5878 | |||
NR_135502.1 | 23 | 5944 | ||||
NR_135503.1 | 23 | 5919 | ||||
NR_135504.1 | 22 | 5701 | ||||
XM_005254910.3 | XP_005254967.1 | 24 | 5901 | |||
XM_011521565.2 | XP_011519867.1 | 18 | 4793 | |||
XM_017022184.1 | XP_016877673.1 | 24 | 5804 | |||
GenBank mRNA | AF116691.1 | - | 1 | 391 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
