siRNA Details
Gene Symbol: AHNAK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138420.2 | NP_612429.2 | 7 | 4337 | ||
XM_005267299.1 | XP_005267356.1 | 7 | 4190 | |||
GenBank mRNA | AB095939.1 | - | 1 | 1672 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
