siRNA Details
Gene Symbol: MOB3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130807.2 | NP_570719.1 | 1, 2 | 55 | ||
GenBank mRNA | AK095471.1 | - | 1, 2 | 55 | ||
AL832474.1 | - | 1, 2 | 91 | |||
CR627409.1 | - | 1, 2 | 56 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
