siRNA Details
Gene Symbol: NLRP2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001319967.1 | NP_001306896.1 | 1 | 1941 | ||
GenBank mRNA | AK026387.1 | - | 2 | 1440 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
