siRNA Details
Gene Symbol: PLXNB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012401.3 | NP_036533.2 | 31, 32 | 5140 | ||
XM_005261909.1 | XP_005261966.1 | 32, 33 | 5188 | |||
XM_005261910.2 | XP_005261967.1 | 32, 33 | 5199 | |||
XM_005261911.4 | XP_005261968.1 | 31, 32 | 5138 | |||
XM_006724413.2 | XP_006724476.1 | 33, 34 | 5258 | |||
XM_011530682.2 | XP_011528984.1 | 32, 33 | 5339 | |||
XM_017028703.1 | XP_016884192.1 | 33, 34 | 5239 | |||
XM_017028704.1 | XP_016884193.1 | 32, 33 | 5192 | |||
GenBank mRNA | AB002313.1 | - | 30, 31 | 5057 | ||
AB075511.1 | - | 16, 17 | 2250 | |||
AK126394.1 | - | 12, 13 | 1849 | |||
BC110366.1 | - | 3, 4 | 377 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
