siRNA Details
Gene Symbol: RPS4X | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007.4 | NP_000998.1 | 7 | 939 | ||
GenBank mRNA | AK222479.1 | - | 7 | 868 | ||
BC000472.2 | - | 6 | 851 | 2961540 | ||
BC007308.1 | - | 6 | 783 | |||
BC071662.1 | - | 6 | 880 | 5505465 | ||
M22146.1 | - | 6 | 848 | |||
M58458.1 | - | 7 | 878 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
