siRNA Details
Gene Symbol: HIST1H2BF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003522.3 | NP_003513.1 | 1 | 374 | ||
GenBank mRNA | BC096120.1 | - | 1 | 374 | 40002352 | |
BC096123.1 | - | 1 | 374 | 40002357 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
